
2-Phenylmorpholine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23972-42-1
:
Formula:C₁₀H₁₃NO·ClH
MDL Number:MFCD03869024
Melting Pt:148…151 °C
:199.677 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together