
2-(4-Chlorophenyl)-2-phenylethylamine hydrochloride
Catalog # APOSOR12744-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:268.182 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:21998-50-5
Formula:C₁₄H₁₄ClN·ClH
MDL Number:MFCD02089402