Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
2-(2-Chlorophenyl)-2-(morpholin-4-yl)ethylamine 95%
Catalog # APOSOR12703-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2-Chlorophenyl)-2-(morpholin-4-yl)ethylamine 95%
Catalog # APOSOR12703-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.729 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:866782-01-6
Formula:C₁₂H₁₇ClN₂O
MDL Number:MFCD01631948