Order Entry
Czech Republic
ContactUsLinkComponent
3-(2-Methoxyphenoxy)benzenesulphonyl chloride
Corrosive
3-(2-Methoxyphenoxy)benzenesulphonyl chloride
Catalog # APOSOR12567-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-(2-Methoxyphenoxy)benzenesulphonyl chloride
Catalog # APOSOR12567-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :298.742 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
 :GHS05
CAS Number:521980-26-7
Formula:C₁₃H₁₁ClO₄S
MDL Number:MFCD01631872