
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916766-91-1
:
Formula:C₁₂H₁₃N₃OS
MDL Number:MFCD09817540
:247.316 g/mol
Melting Pt:94…95 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together