
3-(1-Methyl-1H-pyrazol-5-yl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:941716-85-4
:
Formula:C₁₀H₉ClN₂O₂S
MDL Number:MFCD09817501
:256.709 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together