

1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
Catalog # APOSOR12421-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:231.079 g/mol
Melting Pt:227.5…231.5 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
:GHS07
CAS Number:906352-57-6
Formula:C₉H₇ClN₂O·ClH
MDL Number:MFCD09817460