Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Pyrimidin-2-yl)benzenesulphonyl chloride
4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%
4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%
4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%
APOSOR12391-250MG
:
Apollo Scientific
:
4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%
APOSOR12391-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
254.693 g/mol
:
159.5…161.5 °C
:
Ambient
:
3261
:
8,III
:
GHS05
:
912569-53-0
:
C₁₀H₇ClN₂O₂S
:
MFCD09817488