Order Entry
Czech Republic
ContactUsLinkComponent
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid 97%
Toxic
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid 97%
Catalog # APOSOR12221-5G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid 97%
Catalog # APOSOR12221-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :263.316 g/mol
Melting Pt:247.5…249 °C
 :GHS06
Storage Temperature:Ambient
CAS Number:910037-25-1
Formula:C₁₂H₁₃N₃O₂S
MDL Number:MFCD09065023