Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4-dichlorobenzoic acid
3-Bromo-2,4-dichlorobenzoic acid
3-Bromo-2,4-dichlorobenzoic acid
APOSOR11909-100MG
:
Apollo Scientific
:
3-Bromo-2,4-dichlorobenzoic acid
APOSOR11909-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
269.908 g/mol
:
GHS07
:
Ambient
:
951884-96-1
:
C₇H₃BrCl₂O₂
:
MFCD09800918