1-(Dibenzylamino)propan-2-one
Catalog # APOSOR111617-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.339 g/mol
Storage Temperature:Ambient
CAS Number:68543-42-0
Formula:C₁₇H₁₉NO
MDL Number:MFCD15480865