Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
APOSOR111591-1G
:
Apollo Scientific
:
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
APOSOR111591-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
255.068 g/mol
:
Ambient
:
1016843-27-8
:
C₉H₇BrN₂O₂
:
MFCD09945508