1-(4-Chlorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Catalog # APOSOR111588-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:348.055 g/mol
Storage Temperature:Ambient
CAS Number:394664-62-1
Formula:C₁₄H₁₀Cl₄N₂
MDL Number:MFCD01163929