
2,3-Dichloro-6-methoxyquinoxaline 98%
Catalog # APOSOR11154-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.063 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:39267-04-4
Formula:C₉H₆Cl₂N₂O
MDL Number:MFCD00461063