
2-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Catalog # APOSOR111537-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.178 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:26323-62-6
Formula:C₈H₁₀N₂O
MDL Number:MFCD18427849