Azepan-1-yl[1-(pyrimidin-2-yl)piperidin-3-yl]methanone
Catalog # APOSOR111508-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:288.388 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₂₄N₄O
MDL Number:MFCD31621192