Order Entry
Czech Republic
ContactUsLinkComponent
2-(3,4-Dichlorophenoxy)nicotinonitrile
2-(3,4-Dichlorophenoxy)nicotinonitrile
Catalog # APOSOR111474-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,4-Dichlorophenoxy)nicotinonitrile
Catalog # APOSOR111474-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.095 g/mol
Storage Temperature:Ambient
CAS Number:99902-93-9
Formula:C₁₂H₆Cl₂N₂O
MDL Number:MFCD13238892