Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
APOSOR111451-1G
:
Apollo Scientific
:
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
APOSOR111451-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
225.246 g/mol
:
Ambient
:
265125-69-7
:
C₁₃H₁₁N₃O
:
MFCD01765387