Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
Catalog # APOSOR111449-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
Catalog # APOSOR111449-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.253 g/mol
Storage Temperature:Ambient
CAS Number:1292814-70-0
Formula:C₁₂H₁₅NO₂
MDL Number:MFCD10700294