Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Ethyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Catalog # APOSOR111441-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Catalog # APOSOR111441-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :310.819 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₁ClO₂S₂
MDL Number:MFCD31380598