
2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid
Catalog # APOSOR11143-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.161 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:83863-74-5
Formula:C₈H₉NO₄
MDL Number:MFCD00044904