
6-Chloroimidazo[1,2-a]pyridin-3-amine
Catalog # APOSOR111415-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.596 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:82193-28-0
Formula:C₇H₆ClN₃
MDL Number:MFCD14540214