4-Chloro-2-methylsulphanyl-6-phenylpyrimidine
Catalog # APOSOR111410-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:236.721 g/mol
Storage Temperature:Ambient
CAS Number:91063-60-4
Formula:C₁₁H₉ClN₂S
MDL Number:MFCD07656565