Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3-[5-(1,3-oxazol-5-yl)-2-furyl]thiophene-2-carboxylate
Methyl 3-[5-(1,3-oxazol-5-yl)-2-furyl]thiophene-2-carboxylate
Catalog # APOSOR111272-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-[5-(1,3-oxazol-5-yl)-2-furyl]thiophene-2-carboxylate
Catalog # APOSOR111272-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.28 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₉NO₄S
MDL Number:MFCD30481182