Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR111270-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR111270-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.089 g/mol
Storage Temperature:Ambient
CAS Number:1431965-03-5
 :444791-55-3
Formula:C₁₀H₈Cl₂N₂O₂
MDL Number:MFCD02325395