Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 4-[2-cyano-3-(dimethylamino)prop-2-enoyl]piperazine-1-carboxylate
tert-Butyl 4-[2-cyano-3-(dimethylamino)prop-2-enoyl]piperazine-1-carboxylate
Catalog # APOSOR111258-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-[2-cyano-3-(dimethylamino)prop-2-enoyl]piperazine-1-carboxylate
Catalog # APOSOR111258-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :308.376 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₂₄N₄O₃
MDL Number:MFCD30188175