6-Chloro-N'-[(2-chlorophenyl)methylene]imidazo[1,2-a]pyridine-2-carbohydrazide
Catalog # APOSOR111244-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:333.172 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀Cl₂N₄O
MDL Number:MFCD30188172