6-Chloro-N'-[(4-methoxyphenyl)methylene]imidazo[1,2-a]pyridine-2-carbohydrazide
Catalog # APOSOR111243-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:328.753 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₃ClN₄O₂
MDL Number:MFCD30188171