Order Entry
Czech Republic
ContactUsLinkComponent
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-methylthiourea
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-methylthiourea
Catalog # APOSOR111242-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-methylthiourea
Catalog # APOSOR111242-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.74 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₃ClN₂O₂S
MDL Number:MFCD00158734