Methyl 4-(dicyanomethyl)benzoate
Catalog # APOSOR111209-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:200.193 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₈N₂O₂
MDL Number:MFCD300453