
9H-Fluoren-9-ylmethyl 2-hydroxy-1,1-dimethylethylcarbamate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1187667-02-2
:
Formula:C₁₉H₂₁NO₃
MDL Number:MFCD03788642
:311.375 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together