4-Pyridin-3-yl-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
Catalog # APOSOR111060-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.331 g/mol
Storage Temperature:Ambient
CAS Number:1858250-40-4
Formula:C₁₂H₁₄N₄OS
MDL Number:MFCD28148234