N-Pyridin-3-yl-2-(tetrahydro-2H-pyran-4-ylcarbonyl)hydrazinecarbothioamide
Catalog # APOSOR111059-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.346 g/mol
Storage Temperature:Ambient
CAS Number:1858249-71-4
Formula:C₁₂H₁₆N₄O₂S
MDL Number:MFCD28148235