N-(2,4-Dichloro-6-methylphenyl)morpholine-4-carbothioamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858249-75-8
:
Formula:C₁₂H₁₄Cl₂N₂OS
MDL Number:MFCD28148250
:305.223 g/mol
Storage Temperature:Ambient
Frequently Bought Together