3-(Dimethylamino)-1-(6-methoxy-2-naphthyl)prop-2-en-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1477476-75-7
:
Formula:C₁₆H₁₇NO₂
MDL Number:MFCD00662533
:255.312 g/mol
Storage Temperature:Ambient
Frequently Bought Together