Order Entry
Czech Republic
ContactUsLinkComponent
Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one
Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one
Catalog # APOSOR111011-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one
Catalog # APOSOR111011-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.22 g/mol
Storage Temperature:Ambient
CAS Number:55115-79-2
Formula:C₉H₅N₃OS
MDL Number:MFCD01312791