Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Bis(3,4-dichlorophenyl)urea
Irritant
Environment
1,3-Bis(3,4-dichlorophenyl)urea
Katalogové číslo APOSOR11101-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
1,3-Bis(3,4-dichlorophenyl)urea
Katalogové číslo APOSOR11101-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :350.027 g/mol
Melting Pt:281…282 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:4300-43-0
Formula:C₁₃H₈Cl₄N₂O
MDL Number:MFCD00171464