Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
APOSOR110990-1G
:
Apollo Scientific
:
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
APOSOR110990-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.259 g/mol
:
Ambient
:
1823927-61-2
:
C₅H₈N₂O₂S₂
:
MFCD27995613