Order Entry
Czech Republic
ContactUsLinkComponent
1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid
1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSOR110988-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSOR110988-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :302.348 g/mol
Storage Temperature:Ambient
CAS Number:1858257-28-9
Formula:C₁₅H₁₄N₂O₃S
MDL Number:MFCD26939172