
N-(2,6-Dichlorobenzyl)-N-methylamine
Catalog # APOSOR110982-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.07 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
CAS Number:15205-19-3
Formula:C₈H₉Cl₂N
MDL Number:MFCD04532390