
3-Bromo-5-methylimidazo[1,2-a]pyridine
Catalog # APOSOR110936-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.059 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:5857-47-6
Formula:C₈H₇BrN₂
MDL Number:MFCD19441944