6,7-Dihydro-4H-thieno[3,2-c]pyran-2-ylmethanol
Catalog # APOSOR110925-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:170.229 g/mol
Storage Temperature:Ambient
CAS Number:1265897-62-8
Formula:C₈H₁₀O₂S
MDL Number:MFCD17169813