Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Methyl 6,7-dihydro-4H-thieno[3,2-c]pyran-2-carboxylate
Catalog # APOSOR110919-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 6,7-dihydro-4H-thieno[3,2-c]pyran-2-carboxylate
Catalog # APOSOR110919-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.239 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1823277-91-3
Formula:C₉H₁₀O₃S
MDL Number:MFCD27995615