Order Entry
Czech Republic
ContactUsLinkComponent
5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
Catalog # APOSOR110889-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(4,5,6,7-Tetrahydro-1-benzothien-2-yl)-1,3,4-thiadiazol-2-amine
Catalog # APOSOR110889-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.344 g/mol
Storage Temperature:Ambient
CAS Number:100988-06-5
Formula:C₁₀H₁₁N₃S₂
MDL Number:MFCD11137540