Order Entry
Czech Republic
ContactUsLinkComponent
2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
Catalog # APOSOR110855-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
Catalog # APOSOR110855-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.29 g/mol
Storage Temperature:Ambient
CAS Number:400878-25-3
Formula:C₁₁H₁₃NO₄S
MDL Number:MFCD03001215