
5-Bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1227954-90-6
:
Formula:C₉H₇BrN₂O₂
MDL Number:MFCD12756244
:255.068 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together