Order Entry
Czech Republic
ContactUsLinkComponent
N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
Catalog # APOSOR110739-1G
Supplier:  Apollo Scientific
CAS Number:  
N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
Catalog # APOSOR110739-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.282 g/mol
Storage Temperature:Ambient
CAS Number:1388191-93-2
Formula:C₁₂H₁₈N₂O₄
MDL Number:MFCD16619603