Ethyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Catalog # APOSOR110671-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.274 g/mol
Storage Temperature:Ambient
CAS Number:39067-28-2
Formula:C₁₁H₁₀N₂O₂S
MDL Number:MFCD00052302