Order Entry
Czech Republic
ContactUsLinkComponent
5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
Catalog # APOSOR110644-1G
Supplier:  Apollo Scientific
CAS Number:  
5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
Catalog # APOSOR110644-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.241 g/mol
Storage Temperature:Ambient
CAS Number:1427460-46-5
Formula:C₁₁H₁₅NO₄
MDL Number:MFCD18804159