Order Entry
Czech Republic
ContactUsLinkComponent
Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Irritant
Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Catalog # APOSOR110636-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Catalog # APOSOR110636-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.288 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1355171-29-7
Formula:C₁₄H₁₅N₃O₂
MDL Number:MFCD18074543